Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

WHEATON® FTA® Nucleic Acid Collection Storage Boxes, DWK Life Sciences

Supplier: DWK Life Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

FTA®
W651611 W651612
10796-640CS 67.72 USD InStock
10796-640 89495-470
WHEATON® FTA® Nucleic Acid Collection Storage Boxes, DWK Life Sciences
Boxes Cryoboxes
Storage boxes provide an ideal method for batching/storing FTA® Nucleic acid collection pouches.

  • For use with small or large FTA® cards
  • Accommodates two rows of 3" FTA® card pouches or one row of 6" FTA® card pouches
  • Removable divider for use with smaller pouches
  • Water repellent box material allows for longer durability
  • For use at RT, 4 °C, –20 °C, or colder

Storage boxes are ideal for nucleic acid collection.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty