Would you like to log in again?
PROSAT® Gowning Room Wipes
Supplier: Contec
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PROSAT®
PSCL0001
10797-040CS 238.3 USD InStock
10797-040
PROSAT® Gowning Room Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Gowning room wipes are made using a strong cellulose/polyester non-woven fabric that is folded for easy pickup and very low in extractable residue.
  • Designed for easy pickup
  • Very low in extractable residue
  • Wipes remain clear and saturated
  • Wipes are always conveniently at hand
Saturated with a blend of 6% isopropyl alcohol and 94% deionized water, these wipes are ideal for glove wipe down during gowning procedures. The wipe is folded for easy pickup. Packaged in refill packs and a dispensing tub to keep the wipes clean and saturated.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty