Would you like to log in again?
EXP®2 Fitting Kits for Stainless Steel Tubing
Supplier: Optimize Technologies
EXP®2
15-19-04958 15-20-04933 15-19-04959 15-19-04962 15-19-04961 15-20-04924
10798-778PK 265.03 USD InStock
10798-778 10798-776 10798-588 10798-586 10798-584 10798-780
EXP®2 Fitting Kits for Stainless Steel Tubing
Fittings and Connectors
Optimize is proud to introduce EXP2 - the next generation of EXP UHPLC fitting technology. Featuring a small ³/₁₆" hex-head EXP2 nut, the new EXP2 Driver and the original titanium hybrid ferrule, the EXP2 fittings offer hand-tight connections rated to 20000 psi+. The EXP2 nuts are available with 6-32, 6-40 and 10-32 threads and have been scaled down to allow for maximum working room in cramped spaces. The EXP2 Driver is a small, easy to use knurled torque driver which snaps on to the nuts for hand-tightening and loosening. The EXP2 Driver is slotted and can be removed from the nut and tubing after use.
  • Rated to 20000+ psi (1400+ bar)
  • Tested to 30000 psi
  • Auto-adjusting zero-dead-volume connection
  • Intended for many repeat uses
  • Cost efficient with replaceable components
  • Made in the USA
The patented EXP titanium hybrid ferrule integrates the resilience of titanium with the proven sealing ability of PEEK in a superb one-piece design. The hybrid ferrule provides a perfect seal with every connection, yet the swage can be released without tools to adjust to the different port depths of various hardware. This adjustability provides unlimited value through repeated zero-dead-volume connections with any port configuration.
The EXP and EXP2 fittings are the premier adjustable nut and ferrule compression system for extreme high-pressure connections. The key advantages lie at the confluence of simplicity and versatility. With the EXP and EXP2 fitting systems, you no longer have to clip off and replace swaged ferrules or wonder if you are achieving the best possible connection.
Ordering information: Available in Standard (¹/₁₆") and Nano (¹/₃₂") configurations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty