Would you like to log in again?
Rat Tunnel™ Stainless Steel End Caps, Certified, Bio-Serv
Supplier: Bio-Serv
Rat Tunnel™
K3325-ECM K3326-ECM K3326-ECM K3325-ECM
10798-826EA 46.21 USD InStock
10798-826 CA10798-828 10798-828 CA10798-826
Rat Tunnel™ Stainless Steel End Caps, Certified, Bio-Serv
Animal Supplies Animal Environmental Enrichment
Made of high-temp polycarbonate with stainless steel end caps
An excellent enrichment option for toxicology studies. Transparency makes routine cage checks easier and faster. The stainless steel end caps prevent gnawing of polycarbonate. Autoclavable. Cagewasher safe.
Certifications: Certified (Contaminant Screened).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty