Would you like to log in again?
CritiCore Taffeta-Hypalon/CSM Shoe Covers
Supplier: CritiCore Protective Wear
CritiCore
10798-894 10798-886 10798-888 10798-890 10798-958 10798-892
10798-958PK 27 USD InStock
10798-958 10798-888 10798-886 10798-894 10798-892 10798-890
CritiCore Taffeta-Hypalon/CSM Shoe Covers
Workwear
These reusable cleanroom shoe covers are manufactured with a soft, tan CSM (Hypalon) sole and 100% monofilament polyester taffeta uppers.
  • Tunneled elastic at the ankle
  • Color-coded loop for size identification
  • Single snap adjustment on the back
Extremely lightweight shoe covers are available in white only.
Ordering information: Contact your Avantor sales representative for more information.
Packaging: Shoe covers are packaged in pairs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty