Would you like to log in again?
WRIST ACTION SHAKER MODEL FF 220V 50C
Supplier: Burrell Scientific
Shaker^ Wrist Action* Shaker, model 75 FF, The shaker has a built in timer and an adjustment arm that controls the amplitude of shaking from 0 to 15 degrees arc. Shaker operates at 385 Osc/Min. Material Type: Steel/aluminum, Color: Blue/White, HXLXW 12x17.5x17.5in, 220V/50 Hz
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
WRIST ACTION SHAKER MODEL FF 220V 50C 10799-392 075-775-24-36 Each (80lb) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty