Would you like to log in again?
VARIABLE SPEED SHAKER MODEL CC 220V 50C
Supplier: Burrell Scientific
Shaker^ Wrist Action Shaker, Model 95-CC, Variable Speed, Blue/White, Material Type Steel/aluminum, 220V/50 Hz, From 0 to 450 Oscillations per minute ( +/- 10% ), Complies with UL/CSA and CE standards, HXLXW 12x17.5x17.5in, Accommodates 12 vessels, 6 Finger SUREGRIP Clamps included
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VARIABLE SPEED SHAKER MODEL CC 220V 50C 10799-400 075-795-12-36 Each (72lb) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty