Would you like to log in again?
Potassium Acetate 5.0 M, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
MB-042-0500
10799-504EA 268 USD InStock
10799-504
Potassium Acetate 5.0 M, Rockland Immunochemicals
Buffers Biological Buffers
5.0 M Potassium Acetate (untitrated) DEPC Treated
  • 5.0M concentrated stock solution
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
In molecular biology, potassium acetate is used to precipitate dodecyl sulfate (DS) and DS-bound proteins, allowing the removal of proteins from DNA. It is also used as a salt for the ethanol precipitation of DNA. Potassium acetate is used in mixtures applied for tissue preservation, fixation, and mummification. Potassium acetate is used as a catalyst in the production of polyurethanes.
The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Application Note: This product is a 5.0M concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer contains untitrated 5.0 M Potassium Acetate.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water treated with diethyl pyrocarbonate (DEPC).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty