Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AATF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-X55 200-401-X55
10799-518EA 518 USD InStock
10799-518 CA10799-518
Anti-AATF Rabbit Polyclonal Antibody
Antibodies
AATF (apoptosis antagonizing transcription factor) was initially discovered as an interaction partner of ZIP kinase (ZIPK), a member of death-associated protein (DAP) kinase family of pro-apoptotic serine/threonine kinases. AATF is a phosphoprotein containing an acidic region and a putative leucine zipper domain and nuclear localization signal, features which are typical of transcription factors. AATF inhibits the ZIPK-mediated pro-apoptotic pathway and may activate other anti-apoptotic pathways. Recently, it has also been shown to protect neural cells against oxidative damage induced by amyloid b-peptide and to inhibit aberrant production of the beta-peptide by interacting with Par-4 (prostate apoptosis response-4), another pro-apoptotic leucine zipper protein that is associated with neuronal degeneration in Alzheimer's disease (AD), suggesting that AATF may have potential therapeutic applications in both familial and sporadic forms of AD.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunohistochemsitry: 10 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AATF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty