Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACHE Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-232-0100
10799-536EA 498 USD InStock
10799-536
Anti-ACHE Goat Polyclonal Antibody (BAC)
Antibodies
Acetyl Cholinesterase is abundantly expressed in erythrocytes and is part of the type-B carboxylesterase/lipase family. Anti-Acetyl Cholinesterase cancels signal transduction at the neuromuscular junction via hydrolysis of acetylcholine as it is released into the synaptic cleft by the motor neurons. Anti-Acetyl Cholinesterase antibody is ideal for investigators involved in Neuroscience and Neurotransmitter research.

Anti-Acetyl Cholinesterase Biotin Conjugated Antibody has been assayed against 1.0 µg of Acetyl Cholinesterase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:1,000 to 1:2,000 is suggested for this product.

Type: Primary
Antigen: ACHE
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty