Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACIN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X62 600-401-X62
10799-542EA 518 USD InStock
CA10799-542 10799-542
Anti-ACIN1 Rabbit Polyclonal Antibody
Antibodies
Chromatin condensation and nuclear fragmentation (CCNF) is the hallmark of apoptosis. CCNF is triggered by the activation of members of caspase family, caspase activated DNase (CAD/DFF40), and several novel proteins including AIF and CIDE. A new inducer of chromatin condensation was recently identified and designated Acinus (for apoptotic chromatin condensation inducer in the nucleus). Acinus is cleaved by caspase-3 and an additional unknown protease generating a small active peptide p17, which causes chromatin condensation in vitro when it is added to purified nuclei. Acinus also induces apoptotic chromatin condensation in cells. Acinus is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse and designated AcinusL, AcinusS and AcinusS'.

Recommended Dilutions: ELISA: 1:5,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ACIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty