Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AFP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X80 600-401-X80
10799-588EA 518 USD InStock
10799-588 CA10799-588
Anti-AFP Rabbit Polyclonal Antibody
Antibodies
Alpha-fetoprotein (AFP) is a major plasma protein produced by the yolk sac and the liver during fetal development. In adults, AFP expression is often associated with hepatoma or teratoma. However, hereditary persistance of AFP may also be found in individuals with no obvious pathology. AFP is thought to be the fetal counterpart of serum albumin, and the AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. AFP levels in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 10 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty