Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AID Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-G74 600-401-G74
10799-604EA 518 USD InStock
10799-604 CA10799-604
Anti-AID Rabbit Polyclonal Antibody
Antibodies
AID antibody detects human AID. Activation-induced cytidine deaminase (AID) was initially discovered as a homolog of the apolipoprotein B RNA-editing cytidine deaminase 1 (APOBEC1) that showed cytidine deaminase properties in stimulated B cell lines. It is necessary for somatic hypermutation and class switch recombination in B cells, but inappropriate or dysregulated expression AID is often found in tumors and B cell neoplasms. Although it is structurally and functionally similar to the APOBEC proteins, it appears unlikely that AID deaminates dC to dU residues in HIV cDNA as does APOBEC3G. Anti-AID antibodies are ideal for investigators involved in Apolipoprotein and Infectious Disease research.

Recommended Dilutions: Western Blot: 1-2 ug/mL; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: AID
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty