Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AIFM3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X86 600-401-X86
10799-612EA 518 USD InStock
CA10799-612 10799-612
Anti-AIFM3 Rabbit Polyclonal Antibody
Antibodies
Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Recently a protein similar to the apoptosis-inducing factor (AIF) was cloned and designated AIFL (also known as AIFM3). AIFM3 is expressed ubiquitously and is predominantly localized to the inner membranes of mitochondria. Unlike AIF, AIFM3 does not translocate to the nucleus upon induction of apoptosis. However, overexpression of AIFM3, like AIF, induced cytochrome c release from the mitochondria, cleavage of caspase 3, and ultimately apoptosis, indicating AIFM3 induces apoptosis through caspase activation. Multiple isoforms of AIFM3 are known to exist.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AIFM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty