Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKIRIN2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X98 600-401-X98
10799-678EA 518 USD InStock
CA10799-678 10799-678
Anti-AKIRIN2 Rabbit Polyclonal Antibody
Antibodies
The highly conserved, nuclear-localized Akirin1 and Akirin2 proteins critically regulate the transcription of NF-kB-dependent genes and are required for defense against Gram-negative bacteria in the immune deficiency and NF-kB pathways. Akirin1 is dispensable in the mouse, and neither knockout mice nor cells derived from them have obvious distinctive phenotypes. In contrast, Akirin2 is required for development in the mouse and knockout of both Akirin homologs in mice show that Akirin2 is required downstream of toll-like receptor (TLR), TNF-a and IL-1b signaling, and for the production of IL-6. Akirin2 is functionally closer to the single gene in Drosophila, as the homozygous null D. melanogaster Akirin mutants show a similar, mid-to-early embryonic death.

Recommended Dilutions: ELISA: 1:5,000; Western Blot: 0.5 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: AKIRIN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty