Would you like to log in again?
GC Ferrules, Graphite Vespel®, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
0726660 072656 0726549 073105 0726003 2151XX 215204 215208 0726533 215164 0726563 0726564 0726547 215200 0726673 072653 072666 215108 215465 072651 0726701 0726641 1034060 072652 215133 072674 215105 215163 072668 215103 072670 0726548 0726642 072658 0726561 215165 0726640 0726632 215205 215104 0726557 0726522 215168 072673 072664 0726562 072657 0726700 072665 072662
60361-590PK 121.56 USD InStock
60361-590 60361-594 60361-598 60361-596 14226-308 14226-316 14226-314 14226-312 14226-310 60361-582 60361-580 60361-620 60361-586 60361-584 60361-540 60361-616 14226-306 14226-304 60361-614 14226-302 14226-300 60361-618 10799-840 10799-842 14227-496 60361-612 60361-578 60361-610 60361-606 60361-604 14226-334 60361-608 14226-332 14226-330 14226-298 14226-296 14226-292 60362-668 60361-602 14226-318 60361-600 14226-328 14226-326 14226-324 60362-670 14226-288 14226-366 14226-322 14226-286 14226-320
GC Ferrules, Graphite Vespel®, Trajan Scientific and Medical
Fittings and Connectors
Your chromatography analysis does not end with the selection of the GC column
  • Composite of graphite and Vespel®
  • Mechanically robust
  • Hard material
  • Long lifetime
  • Forms a strong grip with capillary column
These GC ferrules should be used with MS interfaces.
The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: A variety of hole sizes are available for 1/16", 1/8" and 1/4" fittings. Ferrules have a 350°C temperature limit. A variety of profiles are available, including short, blank, 2 and 3-hole ferrules. Ferrules are sold in packs of 10.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty