Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ADCY3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-W91 600-401-W91
10799-872EA 523 USD InStock
CA10799-872 10799-872
Anti-ADCY3 Rabbit Polyclonal Antibody
Antibodies
Adenylate cyclase is the enzyme which produces the “second messenger” signaling molecule, cAMP from ATP. Type III adenylate cyclase is localized to the membranes surrounding neuronal cilia. Much is currently unknown about the function of primary cilia in vertebrates, however, recent work has begun to explore their role in neuronal signaling and neurogenesis. Adenylate Cyclase III Antibody is ideal for researchers interested in cell signaling and neuroscience research.

Recommended Dilutions: ELISA: 1:20,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:2000;

Type: Primary
Antigen: ADCY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty