Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AKT3 Mouse Monoclonal Antibody [clone: 9A5.H9.G7]

Supplier: Rockland Immunochemicals
K-J36 K-J36
10799-946EA 179 USD InStock
10799-946 CA10799-946
Anti-AKT3 Mouse Monoclonal Antibody [clone: 9A5.H9.G7]
Antibodies
Combo Pack: This primary and secondary antibody pair comes with matched antibody pairs to detect and quantify endogenous protein levels of human AKT3. AKT3 Antibody detects AKT3 which is a component of the PI-3 kinase pathway and is activated by phosphorylation at Ser 473 and Thr 308. AKT is a cytoplasmic protein also known as Protein Kinase B (PKB) and rac (related to A and C kinases). AKT is a key regulator of many signal transduction pathways. AKT Exhibits tight control over cell proliferation and cell viability. Overexpression or inappropriate activation of AKT is noted in many types of cancer. AKT mediates many of the downstream events of PI 3-kinase (a lipid kinase activated by growth factors, cytokines and insulin). PI 3-kinase recruits AKT to the membrane, where it is activated by PDK1 phosphorylation. Once phosphorylated, AKT dissociates from the membrane and phosphorylates targets in the cytoplasm and the cell nucleus. AKT has two main roles: (i) inhibition of apoptosis; (ii) promotion of proliferation. Anti-AKT3 Antibody is ideal for investigators involved in Cell Signaling, Neuroscience and Signal Transduction research.

Recommended Dilutions: ELISA: 1:2,000 - 1: 10,000; Flow Cytometry: User Optimized; Immunohistochemsitry: 20 µg/mL; Western Blot: 1:500-1:2000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 9A5.H9.G7
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty