Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-A2M Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-207S 200-106-207S
10799-978EA 148 USD InStock
10799-978 CA10799-978
Anti-A2M Goat Polyclonal Antibody (Biotin)
Antibodies
Anti-alpha-2-Macroglobulin antibody is secreted into the plasma and belongs to the protease inhibitor I39 family. Anti-alpha-2-Macroglobulin inhibits all proteinases utilizing a “bait region,” or specific cleavage site, which traps the proteinase, after which a thioester bond is hydrolyzed and regulates the covalent binding of the protein to proteinase. Anti-alpha-2-Macroglobulin antibody is ideal for investigators interested in Cardiovascular , Protease Inhibitor, or Cell Biology research.

Recommended Dilutions: ELISA: 1:20,000 - 1:100,000; Western Blot: 1:2,000 - 1:10,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty