Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANDVsMgp1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y11 600-401-Y11
10799-998EA 518 USD InStock
CA10799-998 10799-998
Anti-ANDVsMgp1 Rabbit Polyclonal Antibody
Antibodies
Andes virus (ANDV) is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS) (1). It was isolated from an infected Oligoryzomys longicaudatus rodent in Chile, and is highly similar to the Sin Nombre virus (SNV).

Recommended Dilutions: ELISA: User Optimized ; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ANDVsMgp1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Virus, Hantavirus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty