Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABC3735 Goat Polyclonal Antibody (Biotin)

Supplier: Rockland Immunochemicals
200-106-092S 200-106-092S
10800-038EA 148 USD InStock
10800-038 CA10800-038
Anti-ABC3735 Goat Polyclonal Antibody (Biotin)
Antibodies
Uricase is a peroxisomal enzyme that catalyzes the conversion of urea to allantoin. Uricase performs this action by cleaving the purine ring of uric acid rendering it much more soluble within the body for excretion. Uricase within the Bacillus species is of high importance due to its high activity and thermostability over a wide range of pH's. Anti-Uricase Antibody is ideal for investigators in Cell Biology and Microbiology research.

Recommended Dilutions: ELISA: 1:1,000 - 1:2,500; Immunoprecipitation: 1:100; Western Blot: 1:500 - 1:1,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: ABC3735
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype:
Reactivity: Bacillus Species

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty