Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APBA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y29 600-401-Y29
10800-054EA 518 USD InStock
10800-054 CA10800-054
Anti-APBA1 Rabbit Polyclonal Antibody
Antibodies
The Amyloid beta 4 (A4) precursor protein-binding family A member 1 (APBA1) is a member of the X11 protein family and a neuronal adapter protein that interacts with the Alzheimer's disease amyloid precursor protein (APP). APBA1 stabilizes APP and inhibits production of proteolytic APP fragments including the Ab peptide that is deposited in the brains of Alzheimer's disease patients. It is thought to be involved in signal transduction processes and is a putative vesicular trafficking protein in the brain that can form a complex with the potential to couple synaptic vesicle exocytosis to neuronal cell adhesion. APBA1 can also bind to CASK, a multidomain scaffolding protein involved in brain development and synapse formation.

Recommended Dilutions: ELISA: 1:5,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: APBA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty