Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ANAPC10 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y31 600-401-Y31
10800-058EA 518 USD InStock
CA10800-058 10800-058
Anti-ANAPC10 Rabbit Polyclonal Antibody
Antibodies
Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC10 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. It has been suggested that APC10 plays a role to regulate the binding of specific substrates to the APC/C complex, similar to that of coactivators.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ANAPC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty