Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-APOBEC3B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y46 600-401-Y46
10800-074EA 518 USD InStock
10800-074 CA10800-074
Anti-APOBEC3B Rabbit Polyclonal Antibody
Antibodies
The Apolipoprotein B mRNA-editing, enzyme-catalytic, polypeptide-like (APOBEC) 3 is a multi-isoform member of the cytosine deaminase family of enzymes that act on monomeric nucleoside and nucleotide substrates (1). Similar to TRIM5? which targets incoming retroviral capsids, APOBEC3 plays a major role in cellular defense against retroviral infection (2,3). APOBEC3B has also been shown to be upregulated in several cancers; it is thought that APOBEC3B-catalyzed genomic uracil lesions are responsible for a large proportion of mutations in multiple distinct cancers (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: APOBEC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty