Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATXN1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S65-37]

Supplier: Rockland Immunochemicals
200-302-W87 200-302-W87
10800-130EA 539 USD InStock
10800-130 CA10800-130
Anti-ATXN1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S65-37]
Antibodies
Ataxin-1 is a member of the ATXN1 protein family and contains a single AXH domain. It is a neurodegenerative disorder protein thought to have a role in the metabolism of RNA as it has been shown to localize to the RNA and transcription dependent inclusions within the nucleus. A mutation of Ataxin-1 is the cause of spinocerebellar ataxia type-1 (SCA1), a progressive, neurodegenerative disease that is autosomal dominant and primarily affects the Purjinke cells found in brain stem neuronal populations and the cerebellum. Expression of Ataxin-1 is almost ubiquitous, except in the brain where it is isolated to populations of neurons. Anti-Ataxin-1 is ideal for research in Neuroscience.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: ATXN1
Clonality: Monoclonal
Clone: S65-37
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Center
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty