Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATG9A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y88 600-401-Y88
10800-220EA 518 USD InStock
10800-220 CA10800-220
Anti-ATG9A Rabbit Polyclonal Antibody
Antibodies
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components. This process is negatively regulated by TOR (Target of rapamycin) through phosphorylation of autophagy protein APG1. ATG9A, a multi-spanning membrane protein localizing to the Golgi apparatus and late endosomes, has been proposed to mediate membrane transport to generate autophagosomes. ATG9A has also been implicated as a regulator of STING (stimulator of interferon genes)-mediated innate immune response.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 ?g/mL; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ATG9A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty