Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ATRIP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-W95 600-401-W95
10800-242EA 518 USD InStock
10800-242 CA10800-242
Anti-ATRIP Rabbit Polyclonal Antibody
Antibodies
ATRIP, ATR interacting protein, binds to ATR to regulate ATR expression, and is an essential component of the DNA damage checkpoint pathway (Cortez et al, 2001). ATR is recruited to DNA lesions in part through its association with ATRIP, which in turn interacts with the single-stranded DNA binding protein RPA (Ball et al, 2007). DNA replication forks may stall as a result of DNA damage causing phosphorylation of several proteins, including BRCA1 when colocalizing with ATR/ATRIP complex and RPA (Venere et al, 2007). The DNA replication fork stall coincides with BRCA1 directly phosphorylating ATRIP at ser239. ATRIP pS239 Antibody is ideal for researchers interested in cancer research.

Recommended Dilutions: ELISA: 1:5,000; Western Blot: 1:500;

Type: Primary
Antigen: ATRIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Mouse, Non-Human Primates, Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty