Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z00 600-401-Z00
10800-248EA 518 USD InStock
10800-248 CA10800-248
Anti-HA Rabbit Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA (1). Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found (2). These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. H7N9 bird flu is the newest atypical influenza virus infection that has just been reported since early 2013. The emergence of this new strain occurred in China and has become the present focus for possible worldwide pandemic (3).

Recommended Dilutions: ELISA: User Optimized ; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Virus, H7N9 Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty