Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Avian Influenza Neuraminidase Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-G81 600-401-G81
10800-276EA 518 USD InStock
10800-276 CA10800-276
Anti-Avian Influenza Neuraminidase Rabbit Polyclonal Antibody
Antibodies
Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. There was some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. Although it has been known that cleavage site and glycosylation patterns of the HA protein play important roles in determining the pathogenicity of H5 avian influenza viruses, it has only recently been shown that an additional glycosylation site within the globular head of the NA protein also contributes to the high virulence of the H5N1 virus. Anti-Avian Influenza Neuraminidase antibodies are ideal for investigators involved in infectious disease reseach.

Recommended Dilutions: ELISA: 1 ug/mL ; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: Avian Influenza Neuraminidase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Virus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty