Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B2M Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z16
10800-292EA 518 USD InStock
10800-292
Anti-B2M Rabbit Polyclonal Antibody
Antibodies
Beta2-microglobulin (B2M) is a principal component of the Major Histocompatibility Complex (MHC) class I molecule, a ternary membrane protein complex that displays fragments derived from proteolyzed cytosolic proteins on the surface of cells for recognition by the surveillance immune system (1,2). B2M is involved in the presentation of peptide antigens to the immune system and plays a critically important role in immune system function (3). It is expressed on nearly all nucleated cells and contains one Ig-like C1-type (immunoglobulin-like) domain (2,3). Mutations in the Beta 2-microglobulin gene can enhance the progression of malignant melanoma and osteoarthropathy (4,5).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty