Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCL6 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z45
10800-308EA 518 USD InStock
10800-308
Anti-BCL6 Rabbit Polyclonal Antibody
Antibodies
BCL6 was first identified as a proto-oncogene in diffuse large B cell lymphoma. BCL6, a transcriptional repressor, plays an important role in lymphomagenesis. It binds Stat recognition-like DNA elements and is required for affinity maturation of mature B cells in germinal centers and Th1/Th2 differentiation, a process that critically depends on BCL6-mediated transcriptional repression of p53. BCL6 is a pharmacological target for eradication of leukemia-initiating cells in chronic myeloid leukemia (CML).

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty