Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BECN2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z52
10800-318EA 518 USD InStock
10800-318
Anti-BECN2 Rabbit Polyclonal Antibody
Antibodies
Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells (1,2). Beclin 2 is a mammalian specific homolog of the autophagy protein Beclin 1 (3). Like Beclin 1, Beclin 2 interacts with Bcl-2 and class III PI3K complex components. However, Beclin 2 functions in an additional lysosomal degradation pathway and is required for ligand-induced endolysosomal degradation of several G protein-coupled receptors (3). Beclin 2 is also required for agonist-induced lysosome-mediated degradation of EGFR in lung cancer cells, suggesting that it may also play a role in regulating other intracellular signaling pathways (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BECN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty