Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-B2M Rabbit Polyclonal Antibody (HRP)

Supplier: Rockland Immunochemicals
200-403-141-0100
10800-330EA 498 USD InStock
10800-330
Anti-B2M Rabbit Polyclonal Antibody (HRP)
Antibodies
Anti-beta-2-Microglobulin Antibody detects beta-2-Microglobulin. Beta-2-microglobulin is a component of the class I major histocompatibility complex (MHC), which are present on all nucleated cells (excludes red blood cells). It is involved in the presentation of peptide antigens to the immune system. Beta-2-microglobulin associates not only with the alpha chain of MHC class I molecules, but also with class I-like molecules such as CD1 and Qa. Defects in B2M are the cause of hypercatabolic hypoproteinemia. Anti-beta-2-Microglobulin Antibody is ideal for investigators involved in Cell Signaling, Immunology and Cell Biology research.

Anti-beta-2-Microglobulin antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Type: Primary
Antigen: B2M
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty