Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BLRO_MYRVE Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-212-0100
10800-368EA 498 USD InStock
10800-368
Anti-BLRO_MYRVE Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Bilirubin Oxidase antibody is an enzyme that increases the oxidation rate of bilirubin to biliverdin and other substrates. It is ideal for investigators interested in Metabolism or Signal Transduction research.

Anti-Bilirubin Oxidase has been assayed against 1.0 ug of Blilirubin Oxidase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:1,000 to 1:3,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: BLRO_MYRVE
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty