Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Blocking Buffer for Fluorescent Western Blotting, Rockland Immunochemicals

Supplier: Rockland Immunochemicals
MB-070-0050 MB-070-010TF MB-070S
10800-384EA 73 USD InStock
10800-384 10800-386 10800-388
Blocking Buffer for Fluorescent Western Blotting, Rockland Immunochemicals
Buffers Electrophoresis Buffers Blocking Buffers
This blocking buffer is specifically formulated to achieve superior reproducible western blotting images using fluorescent systems.

  • Concentrated solution
  • Fluorescence technology is widely used to detect proteins.
  • However, many common visible fluorophores often result in considerable background fluorescence in the visible range.

Visible fluorophores are rarely used for membrane-based protein detection because of this high background.  IRDye™800, IRDye™700DX, Alexa Fluor® 680 and Cy5.5™ antibody and reagent conjugates are specifically designed for protein detection methods that use longer-wavelength, near-infrared (IR) fluorophores to visualize proteins in western blotting and other applications.  IRDye™800 and IRDye™700DX fluoresce outside the range of most autofluorescence and therefore specific signal is sharply evident from any background giving the best possible signal-to-noise ratio.  Detection levels in the picogram range on Western blots rival the sensitivity of chemiluminescence on film.  IRDye™800 conjugates are also suitable for immunofluorescence microscopy using commercially available excitation/emission filters in the 780nm/820nm range.  Dual simultaneous labeling in western blots or microscopy is achieved when IRDye™800 conjugates are used in conjunction with IRDye™700 or Cy5.5™ conjugates. IRDye™800 and IRDye™700DX conjugates provide an ultra-sensitive and convenient alternative to standard chemiluminescent protein detection methods, as well as a valuable tool for multicolor imaging. Once reacted with the membrane and dried, IRDye™800 and IRDye™700DX conjugated antibody-protein complexes are very stable, and membranes can be stored protected from light, re-washed and/or rescanned. 

Application Note: Fluorescence technology is widely used to detect proteins in both the visible and near-infrared ranges. This product allows for superior signal detection and lower background noise when fluorochrome conjugated antibodies are used to visualize proteins in western blotting and other applications. Antibody conjugates prepared with IRDye® 800 and IRDye® 700DX (Licor), Cy2™, Cy3™, Cy3.5™, Cy5™ and Cy5.5™ (GE Healthcare), DyLight™405, DyLight™ 549, DyLight™ 649, DyLight™ 680, and DyLight™ 800 (Thermo Fisher/Pierce) and Alexa Fluor® 488, Alexa Fluor® 532, Alexa Fluor® 546, Alexa Fluor® 647 and Alexa Fluor® 680 (Invitrogen/Molecular Probes) have been validated on various platforms using this product with superior results compared to other commercially available products. In the infrared range, where little to no autofluorescence occurs, specific signal is sharply evident from any background giving the best possible signal-to-noise ratio. This allows for detection levels in the picogram range which rivals the sensitivity of chemiluminescence on film for western blotting. Superior results are also seen when this product is used for simultaneous labeling (multiplex) in western blots or microscopy using various fluorochrome combinations for multicolor imaging. Membranes blocked with this product can be dried and are very stable. Membranes that are stored protected from light can be re-washed and/or rescanned.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty