Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BECN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-G85
10800-396EA 518 USD InStock
10800-396
Anti-BECN1 Rabbit Polyclonal Antibody
Antibodies
BMI-1 antibdoy detects human DMI-1. The transcriptional repressor BMI-1 was first identified as a proto-oncogene frequently activated by Moloney murine leukemia proviral insertions in mice and cooperating with c-myc in the generation of mouse lymphomas. BMI-1 is involved in segment specification, cell growth and maintenance, transcriptional regulation, and chromatin modification. A major target of BMI-1 is the ink4a locus which encodes tumor suppressor proteins p16 and p19Arf, which are important in tumor progression and thought to be critical in cell proliferation and senescence. Recent studies have also shown that BMI-1 is required for the maintenance of adult normal and leukemic stem cells, suggesting that BMI-1 could an attractive therapeutic target for stem cell proliferation and renewal as well as for anti-cancer strategies. Anti-BMI-1 is ideal for investigators involved in chromatin, DNA damage and repair, stem cell, cell cycle protein and cancer research.

Recommended Dilutions: Western Blot: 0.5-1 ug/mL; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: BECN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty