Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]

Supplier: Rockland Immunochemicals
200-301-P24
10800-500EA 528 USD InStock
10800-500
Anti-CALD1 Mouse Monoclonal Antibody [clone: CD-7]
Antibodies
Caldesmon is a potential actomyosin regulatory protein found in smooth muscle and nonmuscle cells. The predicted smooth-muscle polypeptide is 793 amino acids long. The high molecular weight caldesmon (h-CaD) is predominantly expressed in smooth muscles, whereas the low molecular weight caldesmon (l-CaD) is widely distributed in nonmuscle tissues and cells. Hayashi et al. (1992) demonstrated that the human CDM gene is composed of 14 exons. This antibody is suitable for researchers interested in neurodegenerative diseases, developmental signaling, GPCR signaling, and MAK Kinase Signaling.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 1-2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: CD-7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty