Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNN2 Mouse Monoclonal Antibody [clone: CP-19]

Supplier: Rockland Immunochemicals
200-301-P26
10800-510EA 528 USD InStock
10800-510
Anti-CNN2 Mouse Monoclonal Antibody [clone: CP-19]
Antibodies
Calponin is a basic 34-kD protein first isolated from chicken gizzard and bovine aorta. It is specifically expressed in smooth muscle and binds calmodulin, actin, and tropomyosin. It is able to inhibit the ATPase activity of myosin and is thought to play a role in smooth muscle contraction. Calponin is a smooth muscle specific, actin-, tropomyosin- and calmodulin-binding protein thought to be involved in some way in the regulation or modulation of contraction. This antibody is suitable for researchers interested in cardiovascular diseases, Calcium Regulation in Cardiac Cells, myometrial pathways, skin cancer research, and down syntrome.

Recommended Dilutions: ELISA: 1:5000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.25-0.5ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CNN2
Clonality: Monoclonal
Clone: CP-19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty