Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CADPS Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB0
10800-514EA 518 USD InStock
10800-514
Anti-CADPS Rabbit Polyclonal Antibody
Antibodies
CAPS1 and its related protein CAPS2 encode novel neural/endocrine-specific cytosolic and peripheral membrane proteins. Both are essential components of the synaptic vesicle priming machinery and are required for the Ca2+-regulated exocytosis of secretory vesicles; CAPS-deficienct neurons contain no or very few fusion competent synaptic vesicles, causing a selective impairment of fast phasic transmitter release. CAPS1 acts at a stage in exocytosis that follows ATP-dependent priming, which involves the essential synthesis of phosphatidylinositol 4, 5-bisphosphate and is thought to be a specific regulator of large dense-core vesicle fusion.

Recommended Dilutions: ELISA: 1:2500-1:5000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.25-0.5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CADPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty