Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CARM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB7
10800-528EA 518 USD InStock
10800-528
Anti-CARM1 Rabbit Polyclonal Antibody
Antibodies
Protein arginine N-methyltransferases, such as CARM1, catalyze the transfer of a methyl group from S-adenosyl-L-methionine to the side chain nitrogens of arginine residues within proteins to form methylated arginine derivatives and S-adenosyl-L-homocysteine. Protein arginine methylation has been implicated in signal transduction, metabolism of nascent pre-RNA, and transcriptional activation. CARM1 functions as a transcriptional co-activator for various nuclear receptors and NF-kB. It has also been shown to methylate histone H3 arginines, inhibiting the binding of corepressors and protecting chromatin from deacetylation, thereby facilitating transcription.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: User Optimized; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CARM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty