Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CARD11 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AB8
10800-530EA 518 USD InStock
10800-530
Anti-CARD11 Rabbit Polyclonal Antibody
Antibodies
CARMA proteins belong to the membrane-associated guanylate kinase-like (MAGUK) family of proteins that can function as molecular scaffolds that assist assembly of signal transduction molecules. CARMA1, CARMA2, and CARMA3 share high degrees of sequence and functional homology, but their tissue-specific distribution suggests that they serve distinct biological functions in different cell types. Both CARMA1 and CARMA3 associate with NEMO, the regulatory subunit of the IkK complex, thereby regulating activation of the NF-kB transcription factor. Also, gene inactivation studies showed a complete block in T and B cell immunity as well as an impaired response to LPS, indicating that CARMA1 is a critical regulator in both the adaptive and innate immune systems.

Recommended Dilutions: ELISA: 1:5000-1:20,000; Immunohistochemsitry: 10 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 0.5-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CARD11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty