Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AE4
10800-566EA 518 USD InStock
10800-566
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. A novel member in the caspase family was recently identified and designated ICE-LAP6, Mch6, and Apaf-3. Caspase-9 and Apaf-1 bind to each other, which leads to caspase-9 activation. Caspase-9 is also activated by granzyme B and CPP32. Activated caspase-9 cleaves and activates caspase-3 that is one of the key proteases, being responsible for the proteolytic cleavage of many key proteins in apoptosis. Caspase-9 play a central role in cell death induced by a wide variety of apoptosis activators including TNFa, TRAIL, anti-CD-95, FADD, and TRADD. Caspase-9 is expressed in a variety of human tissues.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Immunoprecipitation: User Optimized; Western Blot: 1:1,000; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty