Would you like to log in again?
Cat Gamma Globulin Fraction
Supplier: Rockland Immunochemicals
D601-0050
10800-570EA 339 USD InStock
10800-570
Cat Gamma Globulin Fraction
Antibodies
Lyophilized Gamma Globulin Fraction
  • prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
  • control reagent
  • Western Blotting and ELISA experiments
Gamma globulins are a class of globulins, identified by their position after serum protein electrophoresis. The most significant gamma globulins are immunoglobulins ("Igs"), more commonly known as antibodies, although some Igs are not gamma globulins, and some gamma globulins are not Igs. Injections are also used to boost immunity in patients unable to produce gamma globulins naturally because of an immune deficiency, such as X-linked a gamma globulinemia and hyper IgM syndrome. Cat Gamma Globulin Fraction is ideal for investigators involved in serum protein component research.
CAT Gamma Globulin Fraction is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency. Specific conditions should be optimized by user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty