Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JUP Mouse Monoclonal Antibody [clone: CN-3]

Supplier: Rockland Immunochemicals
200-301-P27
10800-610EA 528 USD InStock
10800-610
Anti-JUP Mouse Monoclonal Antibody [clone: CN-3]
Antibodies
Junction Plakoglobin (JUP) ,also knows as catenin gamma, is a major cytoplasmic protein that occurs in a soluble and a membrane-associated form and is the only known constituent common to the submembranous plaques of both kinds of adhering junctions, the desmosomes and the intermediate junctions. It is a component of the cadherin-catenin complex, which is predominantly localized where actin filaments anchor in adherens junctions of epithelial cells. The human plakoglobin gene localizes on chromosome 17q21. Gamma-catenin is regulated by the APC tumor suppressor and its oncogenic activity is distinct from that of beta-catenin. This antibody is suitable for researchers interested in cell adhesion and cancer research.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 2ug/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: JUP
Clonality: Monoclonal
Clone: CN-3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty