Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDC25C Mouse Monoclonal Antibody [clone: IMD-25]

Supplier: Rockland Immunochemicals
200-301-P28
10800-708EA 528 USD InStock
10800-708
Anti-CDC25C Mouse Monoclonal Antibody [clone: IMD-25]
Antibodies
Cell division sycle(CDC25) gene product is a protein-tyrosine phosphatase, activates a partially purified p34(cdc2)/cyclin B complex. The cdc25 protein also shares homology with a protein phosphatase with activity against both tyrosine and serine (and thus probably threonine) phosphate residues. CDC25C is expressed predominantly in G2 phase in healer cells. The human gene encodes a protein with a predicted molecular mass of 53,000 daltons whose C-terminal domain shares about 37% sequence identity with the fission yeast cdc25 mitotic inducer. CDC25C gene is mapped to 5q31. This antibody is suitable for researchers interested in cancer research, autoimmune disease, cardiovascular disease, neurodegenerative disease, MAK Kinase Signaling, AKT Signaling, GPCR Signaling, Cell Cycle research, and nuclear receptors and transcription factors.

Recommended Dilutions: ELISA: 1:20,000; Western Blot: 1-2µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CDC25C
Clonality: Monoclonal
Clone: IMD-25
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty