Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CFTR Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AH7
10800-774EA 523 USD InStock
10800-774
Anti-CFTR Rabbit Polyclonal Antibody
Antibodies
The cystic fibrosis transmembrane conductance regulator (CFTR) protein is a member of the ATP-binding cassette (ABC) transporter superfamily, and a member of the MRP subfamily that is involved in multi-drug resistance (1,2). CFTR functions as a chloride channel and controls the regulation of other transport pathways (3). Mutations in this gene are associated with the autosomal recessive disorder cystic fibrosis, the most common, fatal, inherited disease of caucasian populations (1).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CFTR
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty