Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-COX4I1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X00
10800-888EA 518 USD InStock
10800-888
Anti-COX4I1 Rabbit Polyclonal Antibody
Antibodies
COX, also known as cytochrome c oxidase, has 12 subunits that make up the transmembrane mitochondrial protein. Subunit IV has two isoforms; COXIV-1 and COXIV-2. COXIV-1 is expressed ubiquitously while COXIV-2 is highly expressed in adult lung and low levels in brain and heart. Phosphorylation of amino acid residue ser58 of the COXIV-1 protein is a PKA-dependent regulation of COX and plays an important role in metabolism and CREB cycle activation. COX IV pS58 Antibody is ideal for researchers interested in cell cycle research.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000;

Type: Primary
Antigen: COX4I1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty