Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KCP Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AN4
10800-908EA 518 USD InStock
10800-908
Anti-KCP Rabbit Polyclonal Antibody
Antibodies
CRIM2, also known as Kielin/chordin-like protein (KCP), is a novel enhancer of BMP signaling. It binds to BMP7, a morphogen that is important for kidney development and part of the kidney's physiological response to repair of acute kidney injury, enhancing its binding to its receptor. Because of this, CRIM2 has been suggested as a potential antifibrotic therapy in acute and chronic kidney injury. CRIM2 can also interact directly with either activin or TGF-b1, suppressing activin- and TGF-b1-dependent transcription.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: KCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty