Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF10A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-AX5
10800-998EA 518 USD InStock
10800-998
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Antibodies
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel death domain containing receptor was recently identified and designated DR4 (for death receptor 4). The ligand for this novel death receptor has been identified and termed TRAIL, which is a new member in the TNF family. DR4 is also called TRAIL receptor-1 (TRAIL-R1). DR4 is expressed in most of human tissues including spleen, peripheral blood leukocytes, small intestine and thymus. Like TNFR1, Fas and DR3, DR4 mediates apoptosis and NF-kB activation in many tissues and cells.

Recommended Dilutions: ELISA: 1:5,000; Western Blot: 0.5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TNFRSF10A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty