Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]

Supplier: Rockland Immunochemicals
200-301-P30
10801-044EA 528 USD InStock
10801-044
Anti-CCNA2 Mouse Monoclonal Antibody [clone: CY-28]
Antibodies
The mammalian A-type cyclin family consists of 2 members, cyclin A1 (CCNA1) and cyclin A2 (CCNA2). Human cyclin A gene is assigned to 4q26-q27. Cyclin A protein is synthesized and localized into the nucleus at the onset of S phase in nontransformed mammalian fibroblasts. This antibody is suitable for researchers interested in cancer research, apoptosis, autoimmune disease, neurodegenerative disease, cardiovascular disease, MAP Kinase Signaling, AKT Signaling, Inflammatory Signaling, and Cell Cycle research.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: CCNA2
Clonality: Monoclonal
Clone: CY-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty